missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASE11 Monoclonal Antibody (OTI9C4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810574S
This item is not returnable.
View return policy
Description
RNASE11 Monoclonal antibody specifically detects RNASE11 in Human samples. It is validated for Western BlotSpecifications
| RNASE11 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H9J2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 65080 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI9C4 | |
| Unconjugated | |
| MRPL44 | |
| C14orf6; epididymis secretory sperm binding protein Li 84p; HEL-S-84p; Probable ribonuclease 11; putative inactive ribonuclease 11; RAJ1; ribonuclease 11; ribonuclease A family member 11 (inactive); ribonuclease A j1; ribonuclease, RNase A family, 11 (non-active); RNase 11; RNASE11; UNQ5832/PRO19669 | |
| Full length human recombinant protein of human MRPL44 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction