Learn More
RNASEH2B Antibody (419), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-43774
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
RNASEH2B Monoclonal specifically detects RNASEH2B in Human samples. It is validated for Western Blot.
Specifications
| RNASEH2B | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| RNASEH2B | |
| Recombinant protein encompassing a sequence within the center region of human RNaseH2B. The exact sequence is proprietary. | |
| Protein G purified | |
| RUO | |
| 79621 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| 419 | |
| Western Blot 1:500-1:3000 | |
| ribonuclease H2, subunit B | |
| Mouse | |
| 35 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |