missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNaseH2B Monoclonal Antibody (3I4)

Description
Recommended positive controls: A431, HeLa, and HepG2 cells. Predicted reactivity: Pig (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RNASEH2B is composed of a single catalytic subunit (A) and two non-catalytic subunits (B and C) and specifically degrades the RNA of RNA:DNA hybrids. RNASEH2B is the non-catalytic B subunit of RNase H2, which is thought to play a role in DNA replication.
Specifications
Specifications
| Antigen | RNaseH2B |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 3I4 |
| Concentration | 1.94 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | RNASEH2B |
| Gene Accession No. | Q5TBB1 |
| Gene Alias | 1110019N06Rik; 2610207P08Rik; AGS2; Aicardi-Goutieres syndrome 2 protein; Deleted in lymphocytic leukemia 8; deleted in lymphocytic leukemia 8 homolog; DLEU8; ribonuclease H2; ribonuclease H2 subunit B; ribonuclease H2, subunit B; ribonuclease HI subunit B; RNase H2 subunit B; Rnaseh2b |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?