missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNaseH2B Monoclonal Antibody (GT479)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531529
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Pig (85%). Positive Control: A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RNASEH2B is composed of a single catalytic subunit (A) and two non-catalytic subunits (B and C) and specifically degrades the RNA of RNA:DNA hybrids. RNASEH2B is the non-catalytic B subunit of RNase H2, which is thought to play a role in DNA replication.
Specifications
| RNaseH2B | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 20% glycerol and no preservative; pH 7 | |
| Q5TBB1 | |
| RNASEH2B | |
| Recombinant protein encompassing a sequence within the center region of human RNaseH2B. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| GT479 | |
| Unconjugated | |
| RNASEH2B | |
| 1110019N06Rik; 2610207P08Rik; AGS2; Aicardi-Goutieres syndrome 2 protein; Deleted in lymphocytic leukemia 8; deleted in lymphocytic leukemia 8 homolog; DLEU8; ribonuclease H2; ribonuclease H2 subunit B; ribonuclease H2, subunit B; ribonuclease HI subunit B; RNase H2 subunit B; Rnaseh2b | |
| Mouse | |
| Protein G | |
| RUO | |
| 79621 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction