missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RND1 Antibody (OTI1C3), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73919PECY55
This item is not returnable.
View return policy
Description
RND1 Monoclonal specifically detects RND1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| RND1 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| RND1 | |
| Full length recombinant protein of human RND1 (NP_055285) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI1C3 | |
| Flow Cytometry | |
| ARHS, GTP-binding protein, ras homolog gene family, member S, Rho family GTPase 1FLJ42294, Rho6, rho-related GTP-binding protein Rho6, RHOS, Rnd1 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 27289.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction