missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RND1 Monoclonal Antibody (OTI1C3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501691S
This item is not returnable.
View return policy
Description
RND1 Monoclonal antibody specifically detects RND1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| RND1 | |
| Monoclonal | |
| 0.36 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NEX9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 121214 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI1C3 | |
| Unconjugated | |
| SDR9C7 | |
| A830014L09Rik; Arhs; GTP-binding protein RHO6; ras homolog gene family, member S; Rho family GTPase 1; Rho6; Rho-related GTP-binding protein Rho6; RHOS; RND1; similar to Rho-related GTP-binding protein Rho6 (Rho family GTPase 1) (Rnd1) | |
| Full length human recombinant protein of human SDR9C7 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction