missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RND1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16314735 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16314735 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16314735 Supplier Invitrogen™ Supplier No. PA595465

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Human RND1 shares 95.2% amino acid (aa) sequence identity with mouse RND1. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: mouse liver tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Lacks intrinsic GTPase activity. Has a low affinity for GDP, and constitutively binds GTP. Controls rearrangements of the actin cytoskeleton. Induces the Rac-dependent neuritic process formation in part by disruption of the cortical actin filaments. Causes the formation of many neuritic processes from the cell body with disruption of the cortical actin filaments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RND1
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene Rnd1
Gene Accession No. Q8BLR7, Q92730
Gene Alias A830014L09Rik; Arhs; GTP-binding protein RHO6; ras homolog gene family, member S; Rho family GTPase 1; Rho6; Rho-related GTP-binding protein Rho6; RHOS; RND1; similar to Rho-related GTP-binding protein Rho6 (Rho family GTPase 1) (Rnd1)
Gene Symbols Rnd1
Host Species Rabbit
Immunogen E.coli-derived human RND1 recombinant protein (Position: K105-C229).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223881, 27289
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.