missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RND3 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98843AF647
This item is not returnable.
View return policy
Description
RND3 Polyclonal antibody specifically detects RND3 in Human samples. It is validated for Western Blot
Specifications
| RND3 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 647 | |
| ARHE, Protein MemB, Rho family GTPase 3member E, RHO8, RHOE, Rho-related GTP-binding protein Rho8, rho-related GTP-binding protein RhoE, Rnd3, small GTP binding protein Rho8 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human RND3. | |
| 0.1 mL | |
| Signal Transduction | |
| 390 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction