missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF11 Monoclonal Antibody (OTI1G7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810976S
This item is not returnable.
View return policy
Description
RNF11 Monoclonal antibody specifically detects RNF11 in Human samples. It is validated for Western BlotSpecifications
| RNF11 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y255 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 27166 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1G7 | |
| Unconjugated | |
| PRELID1 | |
| CGI-123; N4wbp2; NEDD4 WW domain-binding protein 2; RING finger protein 11; Rnf11; Sid 1669; SID1669 | |
| Human recombinant protein fragment corresponding to amino acids 36-219 of human PRELID1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction