missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNF115 Recombinant Rabbit Monoclonal Antibody (K01_1K42)
GREENER_CHOICE

Product Code. 30825429
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30825429 100 μL
30828982 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825429 Supplier Invitrogen™ Supplier No. MA563227

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

RNF115 Recombinant Monoclonal Antibody for Western Blot

ZNF364 contains 1 RING-type zinc finger. The exact function of ZNF364 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF115
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1K42
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene RNF115
Gene Accession No. Q9Y4L5
Gene Alias 2610028E05Rik; AU042696; BCA2; CL469780; E3 ubiquitin-protein ligase RNF115; Rab7-interacting RING finger protein; rabring 7; Rabring7; ring finger protein 115; RING-type E3 ubiquitin transferase RNF115; RNF115; Zfp364; Zinc finger protein 364; ZNF364
Gene Symbols RNF115
Host Species Rabbit
Immunogen A synthetic peptide of human ZNF364.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27246, 362002
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.