missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RNF12 Polyclonal specifically detects RNF12 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | RNF12 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:20 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | DKFZp686N06224, E3 ubiquitin-protein ligase RLIM, E3 ubiquitin-protein ligase RNF12, EC 6.3.2, EC 6.3.2.-, FLJ25923, FLJ41093, LIM domain-interacting RING finger protein, MGC15161, NY-REN-43, Renal carcinoma antigen NY-REN-43, RING finger LIM domain-binding protein, RING finger protein 12FLJ42887, ring finger protein, LIM domain interacting, ring zinc finger LIM domain binding protein, ring zinc finger protein NY-REN-43antigen, R-LIM, RNF12FLJ45986 |
| Gene Symbols | RLIM |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?