missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100319
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RLIM.
RNF12 is a RING-H2 zinc finger protein. It has been shown to be a ubiquitin protein ligase that targets LIM domain binding 1 (LDB1/CLIM), and causes proteasome-dependent degradation of LDB1. This protein and LDB1 are co-repressors of LHX1/LIM-1, a homeodomain transcription factor. Alternatively spliced transcript variants encoding the same protein have been reported.
Specifications
| RNF12 | |
| Polyclonal | |
| Unconjugated | |
| RLIM | |
| AL022832; AW743871; CTD-2530H13.3; DKFZp686N06224; E3 ubiquitin-protein ligase RLIM; E3 ubiquitin-protein ligase RNF12; FLJ25923; FLJ41093; FLJ42887; FLJ45986; Ha1r; Hoxa1 regulated; LIM domain-interacting RING finger protein; MGC15161; NY-REN-43; renal carcinoma antigen NY-REN-43; RING finger LIM domain-binding protein; ring finger protein 12; ring finger protein, LIM domain interacting; ring zinc finger LIM domain binding protein; ring zinc finger protein NY-REN-43antigen; RING-type E3 ubiquitin transferase RLIM; Rlim; R-LIM; Rlim1; RNF12 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 108348168, 19820, 317241, 51132 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9NVW2, Q9WTV7 | |
| RLIM | |
| A synthesized peptide derived from human RLIM(Accession Q9NVW2), corresponding to amino acid residues T310-R360. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction