missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNF121 Monoclonal Antibody (2G7)

Product Code. 15692295
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15692295 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15692295 Supplier Invitrogen™ Supplier No. MA524471

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peptide Sequence: ERDFAEMCAD YMASTIGFYS ESGMPTKHLS DSVCAVCGQQ IFVDVSEEGI IENTYRLSCN HVFHEFCIRG WCIVGKKQTC PYCKEKVDLK RMFSNPWERP HVMYGQLLD.

The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. Several alternatively spliced transcript variants have been noted for this gene, however, not all are likely to encode viable protein products.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF121
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2G7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene RNF121
Gene Accession No. Q9H920
Gene Alias 4930544L10Rik; RING finger protein 121; Rnf121
Gene Symbols RNF121
Host Species Mouse
Immunogen RNF121 (NP_060790.2, 193 a.a. ~ 301 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55298
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.