missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF121 Polyclonal Antibody

RNF121 Polyclonal Antibody
Brand: Invitrogen PA572876
This item is not returnable.
View return policy
Description
RNF121 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
| RNF121 | |
| Polyclonal | |
| Unconjugated | |
| RNF121 | |
| 4930544L10Rik; RING finger protein 121; Rnf121 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 308871, 55298, 75212 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8R1Z9, Q9H920 | |
| RNF121 | |
| RNF121 antibody for a 19 amino acid peptide near the carboxy terminus of human RNF121. The immunogen is within amino acids 260 - 310 of RNF121. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction