missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF122 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-74191
This item is not returnable.
View return policy
Description
RNF122 Polyclonal specifically detects RNF122 in Mouse samples. It is validated for Western Blot.
Specifications
| RNF122 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| RNF122 | |
| Synthetic peptides corresponding to the middle region of Rnf122. Immunizing peptide sequence SLIFCCYFISKLRNQAQSERYGYKEVVLKGDAKKLQLYGTCAVCLEDFKG. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Goat: 100%; Rabbit: 100%; Guinea pig: 92%; Chicken: 85%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| FLJ12526, MGC126622, ring finger protein 122 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79845 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction