Learn More
Invitrogen™ RNF126 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5116912
Product Code. 17229413
  • € 496.00 / 100µL
Description
Positive Samples: HeLa, HepG2, K562, mouse testis, rat liver Immunogen sequence: TGPPPADKEK IQALPTVPVT EEHVGSGLEC PVCKDDYALG ERVRQLPCNH LFHDGCIVPW LEQHDSCPVC RKSLTGQNTA TNPPGLTGVS FSSSSSSSSS S.
The specific function of RNF126 is not yet known. The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNF126
Polyclonal
Unconjugated
RNF126
2610010O19Rik; E3 ubiquitin-protein ligase RNF126; ring finger protein 126; RING finger protein 126; E3 ubiquitin-protein ligase RNF126; RNF126
Rabbit
Affinity Chromatography
RUO
314613, 55658, 70294
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1.2 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Q91YL2, Q9BV68
RNF126
A synthetic peptide corresponding to a sequence within amino acids 200-300 of human RNF126 (NP_9194421).