Learn More
Invitrogen™ RNF133 Monoclonal Antibody (3D6)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA523148
Product Code. 15553192
  • € 520.00 / 100µg
Description
Peptide Sequence: FFHKNCIDPW ILPHGTCPIC KCDILKVLGI QVVVENGTEP LQVLMSNELP ETLSPSEEET NNEVSPAGTS DKVIHVEENP TSQNNDIQPH SVVEDVHPSP.
The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. This gene has no intron.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNF133
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q8WVZ7
Rnf133
RNF133 (NP_631914, 277 a.a. ~ 376 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2b κ
ELISA, Western Blot
3D6
Unconjugated
Rnf133
E3 ubiquitin-protein ligase RNF133; Goliath-related E3 ubiquitin-protein ligase 2; Greul2; RING finger protein 133; RING-type E3 ubiquitin transferase RNF133; Rnf133
Mouse
Affinity Chromatography
RUO
168433
-20°C, Avoid Freeze/Thaw Cycles