missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNF133 Polyclonal Antibody

Product Code. 13425557
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13425557 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13425557 Supplier Invitrogen™ Supplier No. PA531044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: K562. Predicted reactivity: Rhesus Monkey (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. This gene has no intron.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF133
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Rnf133
Gene Accession No. Q8WVZ7
Gene Alias E3 ubiquitin-protein ligase RNF133; Goliath-related E3 ubiquitin-protein ligase 2; Greul2; RING finger protein 133; RING-type E3 ubiquitin transferase RNF133; Rnf133
Gene Symbols Rnf133
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RNF133. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 168433
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.