missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF135 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 292.00 - € 572.00
Specifications
| Antigen | RNF135 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18447620
|
Novus Biologicals
NBP1-81212-25ul |
25ul |
€ 292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18298195
|
Novus Biologicals
NBP1-81212 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RNF135 Polyclonal specifically detects RNF135 in Human samples. It is validated for Western Blot.Specifications
| RNF135 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| E3 ubiquitin-protein ligase RNF135, EC 6.3.2.-, MGC13061, REUL, ring finger protein 135RIG-I E3 ubiquitin ligase, Riplet | |
| RNF135 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Zinc Finger | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 84282 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LSILGKAFSSGVDLSMASPKLVTSDTAAGKIRDILHDLEEIQEKLQESVTWKEAPEAQMQGELLEAPSSSSCPLPDQSHPALRRASRFAQWAIHPTFNLK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title