missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF135 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598596
This item is not returnable.
View return policy
Description
RNF135 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The protein encoded by RNF135 contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. This gene is located in a chromosomal region known to be frequently deleted in patients with neurofibromatosis.
Specifications
| RNF135 | |
| Polyclonal | |
| Unconjugated | |
| RNF135 | |
| 0610037N03Rik; 2410006N06Rik; E3 ubiquitin-protein ligase RNF135; L13; LRRGT00184; MGC13061; MMFD; REUL; RIG-I E3 ubiquitin ligase; ring finger protein 135; RING finger protein 135; E3 ubiquitin-protein ligase RNF135; RING-type E3 ubiquitin transferase RNF135; Riplet; RNF135; U 2-3-0 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84282 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.82 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IUD6 | |
| RNF135 | |
| Recombinant Human E3 ubiquitin-protein ligase RNF135 protein (339-432AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction