missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF14 Monoclonal Antibody (OTI3A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810955
This item is not returnable.
View return policy
Description
The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This protein interacts with androgen receptor (AR) and may function as a coactivator that induces AR target gene expression in prostate. A domit negative mutant of this gene has been demonstrated to inhibit the AR-mediated growth of prostate cancer. This protein also interacts with class III ubiquitin-conjugating enzymes (E2s) and may act as a ubiquitin-ligase (E3) in the ubiquitination of certain nuclear proteins. Five alternatively spliced transcript variants encoding two distinct isoforms have been reported.Specifications
| RNF14 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UBS8 | |
| RNF14 | |
| Human recombit protein fragment corresponding to amino acids 1-220 of human RNF14 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3A4 | |
| Unconjugated | |
| RNF14 | |
| ARA54, HFB30, HRIHFB2038, TRIAD2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9604 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction