missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF141 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540920
This item is not returnable.
View return policy
Description
Peptide sequence: RIMNLYQFIQ LYKDITSQAA GVLAQSSTSE EPDENSSSVT SCQASLWMGR Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 85%; Rat: 93%.
The protein encoded by this gene contains a RING finger, a motif known to be involved in protein-DNA and protein-protein interactions. Abundant expression of this gene was found in the testicular tissue of fertile men, but was not detected in azoospermic patients. Studies of the mouse counterpart suggest that this gene may function as a testis specific transcription factor during spermatogenesis.
Specifications
| RNF141 | |
| Polyclonal | |
| Unconjugated | |
| RNF141 | |
| RFP141; Ring Finger Protein 141; ZFP26; ZNF230 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 50862 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WVD5 | |
| RNF141 | |
| Synthetic peptide directed towards the middle region of human RNF141 (aa 96-145). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction