missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF144A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94282-0.1ml
This item is not returnable.
View return policy
Description
RNF144A Polyclonal antibody specifically detects RNF144A in Human, Rat samples. It is validated for Western Blot
Specifications
| RNF144A | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 6.3.2, EC 6.3.2.-, KIAA0161UBCE7IP4probable E3 ubiquitin-protein ligase RNF144A, ring finger protein 144Aring finger protein 144, RNF144, UbcM4-interacting protein 4, ubiquitin conjugating enzyme 7 interacting protein 4, Ubiquitin-conjugating enzyme 7-interacting protein 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 52-179 of human RNF144A (NP_055561.2). ELLIKEGLETAISCPDAACPKQGHLQENEIECMVAAEIMQRYKKLQFEREVLFDPCRTWCPASTCQAVCQLQDVGLQTPQPVQCKACRMEFCSTCKASWHPGQGCPETMPITFLPGETSAAFKMEEDD | |
| 0.1 mL | |
| Zinc Finger | |
| 9781 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction