Learn More
RNF144B Mouse anti-Human, Clone: OTI11F4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF500699
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
E3 ubiquitin-protein ligase which accepts ubiquitin from E2 ubiquitin-conjugating enzymes UBE2L3 and UBE2L6 in the form of a thioester and then directly transfers the ubiquitin to targeted substrates such as LCMT2, thereby promoting their degradation. Induces apoptosis via a p53/TP53-dependent but caspase-independent mechanism. However, its overexpression also produces a decrease of the ubiquitin-dependent stability of BAX, a pro-apoptotic protein, ultimately leading to protection of cell death; But, it is not an anti-apoptotic protein per se.Specifications
| RNF144B | |
| Monoclonal | |
| Unconjugated | |
| RNF144B | |
| IBRDC2, PIR2, bA528A10.3, p53RFP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 255488 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI11F4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q7Z419 | |
| RNF144B | |
| Human recombit protein fragment corresponding to amino acids 1-256 of human RNF144B produced in E.coli | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.