missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNF148 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15634675
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15634675 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15634675 Supplier Invitrogen™ Supplier No. PA550745

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total RNF148 protein.

RNF148 is a single-pass membrane protein. It contains 1 PA (protease associated) domain and 1 RING-type zinc finger. The exact function of RNF148 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF148
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene RNF148
Gene Accession No. Q8N7C7
Gene Alias ACVRINP1; AIP1; KIAA0705; MGC35222; RING finger protein 148; ring finger protein 148-like; RN148; Rnf148
Gene Symbols RNF148
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human ring finger protein 148.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 378925
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.