missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF148 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542374
This item is not returnable.
View return policy
Description
Peptide sequence: PNSFTRRRSQ IKTDVKKAID QLQLRVLKEG DEELDLNEDN CVVCFDTYKP Sequence homology: Human: 100%; Rat: 85%; Yeast: 82%.
RNF148 is a single-pass membrane protein. It contains 1 PA (protease associated) domain and 1 RING-type zinc finger. The exact function of RNF148 remains unknown.
Specifications
| RNF148 | |
| Polyclonal | |
| Unconjugated | |
| RNF148 | |
| ACVRINP1; AIP1; KIAA0705; MGC35222; RING finger protein 148; ring finger protein 148-like; RN148; Rnf148 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 378925 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N7C7 | |
| RNF148 | |
| Synthetic peptide directed towards the C-terminal of human RNF148 (aa 218-267). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction