missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF151 Polyclonal Antibody
Description
This target displays homology in the following species: Cow: 100%; Dog: 92%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 92%; Rat: 100%.
May be involved in acrosome formation of spermatids.
Specifications
Specifications
| Antigen | RNF151 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | RNF151 |
| Gene Accession No. | Q2KHN1 |
| Gene Alias | 1700010O16Rik; AV047371; LOW QUALITY PROTEIN: RING finger protein 151; ring finger protein 151; RING finger protein 151; LOW QUALITY PROTEIN: RING finger protein 151; Rnf151; Unknown (protein for MGC:133560) |
| Gene Symbols | RNF151 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?