missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF156 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11285-1-AP-20UL
This item is not returnable.
View return policy
Description
Mahogunin (MGRN1) is a C3HC4 RING-containing protein with E3 ubiquitin ligase activity in vitro.Specifications
| RNF156 | |
| Polyclonal | |
| Unconjugated | |
| MGRN1 | |
| KIAA0544, mahogunin, ring finger 1, MGRN1, RING finger protein 156, RNF156 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17237, 23295, 302938 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60291, Q5XIQ4, Q9D074 | |
| Mgrn1 | |
| RNF156 Fusion Protein Ag1808 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction