missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF160 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-57180-25ul
This item is not returnable.
View return policy
Description
RNF160 Polyclonal specifically detects RNF160 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| RNF160 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| C21orf10, C21orf98, E3 ubiquitin-protein ligase listerin, EC 6.3.2.-, KIAA0714ZNF294, listerin E3 ubiquitin protein ligase 1, RING finger protein 160LISTERIN, RNF160FLJ11053, Zinc finger protein 294°Chromosome 21 open reading frame 98 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 26046 | |
| Human | |
| Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| LTN1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ECLRFIMQQNLGEEEIEQMLVNDQLIPFIDAVLKDPGLQHGQLFNHLAETLSSWEAKADTEKDEKTAHNLENVLI | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only