missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF166 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-81203-25ul
This item is not returnable.
View return policy
Description
RNF166 Polyclonal antibody specifically detects RNF166 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| RNF166 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL | |
| MGC14381, MGC2647, ring finger protein 166 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PCRGCNKKVTLAKMRVHISSCLKVQEQMANCPKFVPVVPTSQPIPSNIPNRSTFACPYCGARNLDQQELVKHCVESHRSDPNRVVCPICSAMP | |
| 25 μL | |
| Zinc Finger | |
| 115992 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?