missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF166 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531503
This item is not returnable.
View return policy
Description
Recommended positive controls: U87-MG. Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (93%), Chicken (91%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RNF166 (RING finger protein 166) is a 237 amino acid protein that contains one RING-type zinc finger and a UIM (ubiquitin-interacting motif) repeat. Existing as two alternatively spliced isoforms, RNF166 is encoded by a gene that maps to human chromosome 16q24.3. Chromosome 16 encodes over 900 genes and comprises nearly 3% of the human genome. The GAN gene is located on chromosome 16 and, with mutation, may lead to giant axonal neuropathy, a nervous system disorder characterized by increasing malfunction with growth. The rare disorder Rubinstein-Taybi syndrome is also associated with chromosome 16, as is Crohn's disease, which is a gastrointestinal inflammatory condition.
Specifications
| RNF166 | |
| Polyclonal | |
| Unconjugated | |
| RNF166 | |
| 1110031E24Rik; AW555115; E3 ubiquitin-protein ligase RNF166; RING finger protein 166; RING-type E3 ubiquitin transferase RNF166; RNF166; Zfp313l; zinc finger protein 313; zinc finger protein 228 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 115992 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96A37 | |
| RNF166 | |
| Recombinant protein encompassing a sequence within the center region of human RNF166. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction