missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNF167 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15813114
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15813114 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15813114 Supplier Invitrogen™ Supplier No. PA568184

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

May act as an E3 ubiquitin-protein ligase, or as part of the E3 complex, which accepts ubiquitin from specific E2 ubiquitin-conjugating enzymes, such as UBE2E1, and then transfers it to substrates, such as SLC22A18. May play a role in growth regulation involved in G1/S transition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF167
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RNF167
Gene Accession No. Q5XIL0, Q91XF4, Q9H6Y7
Gene Alias 0610010G05Rik; 5730408C10Rik; AU019305; AV328608; E3 ubiquitin-protein ligase RNF167; LP2254; novel PA domain and C3HC4 type zinc finger (RING finger) containing protein; RGD1305972; RING finger protein 167; RING105; RING-type E3 ubiquitin transferase RNF167; Rnf167
Gene Symbols RNF167
Host Species Rabbit
Immunogen Synthesized peptide derived from human RNF167.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26001, 360554, 70510
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.