missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF168 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20178
This item is not returnable.
View return policy
Description
RNF168 Polyclonal specifically detects RNF168 in Human samples. It is validated for Western Blot.
Specifications
| RNF168 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| E3 ubiquitin-protein ligase RNF168, EC 6.3.2, EC 6.3.2.-, FLJ39749, ring finger protein 168FLJ35794 | |
| Rabbit | |
| 65 kDa | |
| 0.1 mL | |
| Zinc Finger | |
| 165918 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:10000 | |
| Q8IYW5 | |
| RNF168 | |
| Recombinant protein encompassing a sequence within the center region of human RNF168. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction