Learn More
Invitrogen™ RNF181 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589806
Description
Immunogen sequence: MASYFDEHDC EPSDPEQETR TNMLLELARS LFNRMDFEDL GLVVDWDHHL PPPAAKTVVE NLPRTVIRGS QAELKCPVCL LEFEEEETAI EMPCHHLFHS SCILPWLSKT NSCPLCRYEL PTDDDTYEEH RRDKARKQQQ QHRLENLHGA MYT; Positive Samples: LO2, Mouse liver, Mouse brain, Mouse kidney, Mouse lung, Rat testis, Rat brain.
RNF181 binds the integrin alpha-IIb (ITGA2B)/beta-3 (ITGB3) complex and has E3 ubiquitin ligase activity (Brophy et al., 2008).
Specifications
| RNF181 | |
| Polyclonal | |
| Unconjugated | |
| Rnf181 | |
| 1700047F16Rik; 2500002L14Rik; 6230415N01Rik; C77350; E3 ubiquitin-protein ligase RNF181; HSPC238; RING finger protein 181; RING-type E3 ubiquitin transferase RNF181; RNF181 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 297337, 51255, 66510 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.59 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6AXU4, Q9CY62, Q9P0P0 | |
| Rnf181 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-153 of human RNF181 (NP_0575781). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.