missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF185 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 470.00
Specifications
| Antigen | RNF185 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18684251
|
Novus Biologicals
NBP2-93917-0.02ml |
0.02 mL |
€ 188.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18641651
|
Novus Biologicals
NBP2-93917-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RNF185 Polyclonal antibody specifically detects RNF185 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| RNF185 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Zinc Finger | |
| PBS (pH 7.3), 50% glycerol | |
| 91445 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| BSK65-MONO1, BSK65-MONO2, BSK65-PANC1, BSK65-PANC2, BSK65-TEST1, BSK65-TEST2, BSK65-TEST3, FLJ38628, ring finger protein 185 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human RNF185 (NP_689480.2). MASKGPSASASPENSSAGGPSGSSNGAGESGGQDSTFECNICLDTAKDAVISLCGHLFCWPCLHQWLETRPNRQVCPVCKAGISRDKVIPLYGRGSTGQQDPREKTPPRPQGQRPEPENRGGFQGFGFGD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title