Learn More
Invitrogen™ Catalase Recombinant Rabbit Monoclonal Antibody (24GB4290)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557354
 View more versions of this product
Product Code. 30276412
  • € 198.00 / 20µL
Description
Catalase Recombinant Monoclonal Antibody for Western Blot, Flow
Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kDa in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara's disease.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Catalase
Recombinant Monoclonal
Unconjugated
Cat
2210418N07; Cas1; Cas-1; CAT; Cat01; catalase; catalase 1; Catl; CS1; Cs-1; CTA1; HGNC:1516; hypothetical protein LOC531682; MGC138422; MGC138424
Rabbit
Affinity chromatography
RUO
24248, 847
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4290
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P04040, P04762
Cat
Recombinant protein of Mouse Catalase (aa 262-527).
20 μL
Primary
Human, Rat