missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF20 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2242V
This item is not returnable.
View return policy
Description
RNF20 Polyclonal antibody specifically detects RNF20 in Human, Mouse samples. It is validated for Western Blot
Specifications
| RNF20 | |
| Polyclonal | |
| Western Blot | |
| BRE1, BRE1 E3 ubiquitin ligase homolog, BRE1-A, BRE1AMGC129667, E3 ubiquitin-protein ligase BRE1A, EC 6.3.2, EC 6.3.2.-, FLJ11189, FLJ20382, hBRE1MGC129668, homolog of S. cerevisiae BRE1, KAIA2779, ring finger protein 20KIAA2779 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human Ring Finger 20 using the numbering given in entry NP_062538.5 (GeneID 56254). | |
| 0.1 mL | |
| Chromatin Research, Zinc Finger | |
| 56254 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction