missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RNF20 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-27887
This item is not returnable.
View return policy
Description
RNF20 Polyclonal antibody specifically detects RNF20 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| RNF20 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| BRE1, BRE1 E3 ubiquitin ligase homolog, BRE1-A, BRE1AMGC129667, E3 ubiquitin-protein ligase BRE1A, EC 6.3.2, EC 6.3.2.-, FLJ11189, FLJ20382, hBRE1MGC129668, homolog of S. cerevisiae BRE1, KAIA2779, ring finger protein 20KIAA2779 | |
| RNF20 antibody was raised against a 19 amino acid peptide near the amino terminus of human RNF20. The immunogen is located within amino acids 130 - 180 of RNF20. | |
| 0.1 mg | |
| Chromatin Research, Zinc Finger | |
| 56254 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 - 2 μg/mL, ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| Q5VTR2 | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction