missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF20 Monoclonal Antibody (1594CT552.128.36)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537691
This item is not returnable.
View return policy
Description
RNF20 Monoclonal Antibody for Western Blot
RNF20 is an E3 ubiquitin ligase that regulates chromosome structure by monoubiquitinating histone H2B. RNF20 selectively suppresses the expression of some genes such as several protooncogenes and growth-related genes, including many genes that are induced by epidermal growth factor by interfering with chromatin recruitment of transcription elongation factor SII (TFIIS). In contrast, RNF20 also positively regulates the p53 tumor suppressor as well as numerous histone H2A and H2B genes. Together with the deubiquitinase USP44, RNF20 regulates stem cell differentiation by modulating H2B monoubiquitination.
Specifications
| RNF20 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q5VTR2 | |
| RNF20 | |
| recombinant protein of human RNF20. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| 1594CT552.128.36 | |
| Unconjugated | |
| RNF20 | |
| 4833430L21Rik; AW540162; BRE1; BRE1 E3 ubiquitin ligase homolog; BRE1A; BRE1-A; C79397; E3 ubiquitin-protein ligase BRE1A; FLJ11189; FLJ20382; hBRE1; HNF3B; homolog of S. cerevisiae BRE1; hypothetical protein; KIAA2779; Kiaa4116; MGC129667; MGC129668; mKIAA4116; RCJMB04_5a23; RING finger protein 20; ring finger protein 20, E3 ubiquitin protein ligase; RING-type E3 ubiquitin transferase BRE1A; Rnf20; TCF3B | |
| Mouse | |
| Protein G | |
| RUO | |
| 56254 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction