Learn More
RNF212B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562810
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ASTHSLSYRT SSASSGQGIF SFRPSPNGHS GHTRVLTPNN FAQRESTTTL ESLPSFQLPV LQTLYQQRRH MGLPSGR Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 31%.
RNF212B Polyclonal antibody specifically detects RNF212B in Human samples. It is validated for Immunohistochemistry.Specifications
RNF212B | |
Polyclonal | |
Unconjugated | |
RNF212B | |
C14orf164; RING finger protein 212B; RING finger protein C14orf164; RING finger protein C14orf164-like; RNF212B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100507650 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A8MTL3 | |
RNF212B | |
Recombinant protein corresponding to Human RNF212B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only