missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNF217 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15784111 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15784111 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15784111 Supplier Invitrogen™ Supplier No. PA556203

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GQVEIKCPIT ECFEFLEETT VVYNLTHEDS IKYKYFLELG RIDSSTKPCP QCKHFTTFKK Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

RNF217 is an E3 ubiquitin-protein ligase which accepts ubiquitin from E2 ubiquitin-conjugating enzymes in the form of a thioester and then directly transfers the ubiquitin to targeted substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF217
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RNF217
Gene Accession No. Q8TC41
Gene Alias C6orf172; dJ84N20.1; IBR domain containing 1; IBR domain-containing protein 1; IBRDC1; opposite STL; OSTL; Probable E3 ubiquitin-protein ligase RNF217; ring finger protein 217; RNF217
Gene Symbols RNF217
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RNF217. Recombinant protein control fragment (Product #RP-94907).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154214
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.