missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF23 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142040
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognise both reported isoforms (NP_067076.2 and NP_742013.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The function of this protein has not been identified. This gene lies within the major histocompatibility complex class I region on chromosome 6. Alternate splicing results in two transcript variants encoding different isoforms.
Specifications
| RNF23 | |
| Polyclonal | |
| Unconjugated | |
| TRIM39 | |
| 1100001D15Rik; E130103K13Rik; E3 ubiquitin-protein ligase TRIM39; mKIAA4179; RBCC-B30.2; ring finger protein 23; RING-B box-coiled coil-B30.2; RING-B box-coiled-coil-B30.2; RING-type E3 ubiquitin transferase TRIM39; Rnf23; Testis-abundant finger protein; tfp; TRIM39; TRIM39B; tripartite motif containing 39; tripartite motif protein 39; tripartite motif-containing 39; tripartite motif-containing protein 39 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 56658 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9HCM9 | |
| TRIM39 | |
| Peptide with sequence C-NAAPLTIRPPTDWE. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction