missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF25 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18859CL1
This item is not returnable.
View return policy
Description
RNF25 Polyclonal antibody specifically detects RNF25 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| RNF25 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| AO7, EC 6.3.2.-, FLJ13906, ring finger protein 25E3 ubiquitin-protein ligase RNF25, ring finger protein AO7 | |
| Produced in rabbits immunized with E. coli-derived Human RNF25 fragment | |
| 0.1 mL | |
| Apoptosis, Cancer, Lipid and Metabolism, Tumor Suppressors, Zinc Finger | |
| 64320 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction