Learn More
Invitrogen™ RNF3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA540882
Product Code. 15584462
  • € 520.00 / 100µL
Description
Peptide sequence: EVPGDIKGET CSAKQHLDSH RNGETKADDS SNKEAAEEKP EEDNDYHRSD Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 79%; Horse: 79%; Human: 100%; Mouse: 86%; Pig: 86%; Rabbit: 85%; Rat: 86%; Yeast: 75%.
Phosphorylase kinase is a hexadecameric enzyme that is comprised of four copies of four subunits that are encoded by four separate genes: PHKA, PHKB, PHKG, and PH kDa. This serine/threonine specific kinase converts glycogen phosphorylase b to glycogen phosphorylase a, resulting in the release of glucose-1-phophate from glycogen. PHKB (Phosphorylase b kinase regulatory subunit beta) is a 1093 amino acid subunit of phosphorylase kinase that, along with PHKA, has regulatory functions controlled by phosphorylation. Defects in the gene encoding PHKB are the cause of glycogen storage disease type 9B, which is also known as phosphorylase kinase deficiency of liver and muscle. This disease is characterized by a mild phenotype of hepatomegaly with only slightly elevated transaminase and plasma lipids, no clinical muscle involvement, and generally is correlated with a gradual improvement with increasing age. There are four isoforms of PHKB that are produced as a result of alternative splicing events.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNF3
Polyclonal
Unconjugated
PCGF3
2310035N15Rik; AI662857; D630042K08Rik; D6S113E; DADB-17J1.3; DKFZp686D20235; DKFZp686N0336; DONG1; E430039C14; FLJ31942; FLJ36550; FLJ43813; FSH; FSRG1; KIAA9001; MGC129075 protein; MGC129615; MGC40413; NAT; PCGF3; polycomb group ring finger 3; polycomb group RING finger protein 3; ring finger protein 3; RING finger protein 3A; RING3; RNF3; RNF3A
Rabbit
Protein A
RUO
10336
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q3KNV8
PCGF3
Synthetic peptide directed towards the middle region of human PCGF3.