missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF39 Antibody (OTI5E10), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73927R
This item is not returnable.
View return policy
Description
RNF39 Monoclonal specifically detects RNF39 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| RNF39 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RNF39 | |
| Full length recombinant protein of human RNF39 (NP_079512) produced in HEK293T cells. | |
| 0.1 mL | |
| 80352.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI5E10 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| HZF, HZFWLTP (long-term potentiation) induced RING finger protein, LIRFHZFw1, Protein HZFw, ring finger protein 39 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction