missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF39 Monoclonal Antibody (OTI5F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505915S
This item is not returnable.
View return policy
Description
RNF39 Monoclonal antibody specifically detects RNF39 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| RNF39 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H2S5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 80352 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunocytochemistry, Western Blot | |
| OTI5F5 | |
| Unconjugated | |
| Rnf39 | |
| BC038661; DADB-60O11.8; Gm1786; Gm1787; Gm320; HZF; HZFW; Hzfw1; HZFw1 protein; Lirf; LTP (long-term potentiation) induced RING finger protein; LTP-induced RING finger protein; Protein HZFw; RING finger protein 39; RNF39 | |
| Full length human recombinant protein of human RNF39 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction