missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF5 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-04014B
This item is not returnable.
View return policy
Description
RNF5 Polyclonal antibody specifically detects RNF5 in Human samples. It is validated for Western Blot
Specifications
| RNF5 | |
| Polyclonal | |
| Western Blot | |
| EC 6.3.2.-, G16, HsRma1, NG2, Protein G16, ring finger protein 5Ram1 homolog, RING5hsRma1, RMA1E3 ubiquitin-protein ligase RNF5 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human Ring Finger Protein 5 using the numbering given in entry NP_008844.1 (GeneID 6048). | |
| 0.1 mL | |
| Cancer, Cell Biology, Immunology | |
| 6048 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction