missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF70 Antibody (3E10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00064219-M07
This item is not returnable.
View return policy
Description
RNF70 Monoclonal antibody specifically detects RNF70 in Human samples. It is validated for Western Blot, ELISA
Specifications
| RNF70 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| EC 6.3.2, EC 6.3.2.-, FLJ11830, praja 1, praja ring finger 1, PRAJA1, RING finger protein 70, RNF70E3 ubiquitin-protein ligase Praja-1 | |
| PJA1 (NP_660095, 81 a.a. ∽ 180 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DSESSGTNLNIDNEDYSRYPPREYRASGSRRGMAYGHIDSYGADDSEEEGAGPVERPPVRGKTGKFKDDKLYDPEKGARSLAGPPPHFSSFSRDVREERD | |
| 0.1 mg | |
| Lipid and Metabolism, Zinc Finger | |
| 64219 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 3E10 | |
| Western Blot 1:500, ELISA | |
| NP_660095 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction