missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF92 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142048
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000. Preliminary analysis showed no signal in Western Blot.
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to the nucleus and the cytoplasm. Its function has not been identified.
Specifications
| RNF92 | |
| Polyclonal | |
| Unconjugated | |
| TRIM11 | |
| BIA1; E3 ubiquitin-protein ligase TRIM11; Protein BIA1; RING finger protein 92; RING-type E3 ubiquitin transferase TRIM11; RNF92; Trim11; tripartite motif containing 11; tripartite motif protein 11; tripartite motif-containing 11; tripartite motif-containing protein 11 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 81559 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q96F44 | |
| TRIM11 | |
| Peptide with sequence CRPKGGSGDTLAPQ. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction