missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNH1 Monoclonal Antibody (OTI2B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501965S
This item is not returnable.
View return policy
Description
RNH1 Monoclonal antibody specifically detects RNH1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| RNH1 | |
| Monoclonal | |
| 1.02 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BY49 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55825 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2B1 | |
| Unconjugated | |
| Pecr | |
| AW546468; C80305; EGK_05816; hypothetical protein LOC517087; im:7039360; LOC720791; Placental ribonuclease inhibitor; placental RNase inhibitor; PRI; RAI; Ribonuclease inhibitor; ribonuclease/angiogenin inhibitor 1; RNH; Rnh1; si:ch211-213a13.3; testicular tissue protein Li 164; uncharacterized protein rnh1; wu:fd55g05 | |
| Full length human recombinant protein of human PECR produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction